Product Details

SNP ID
rs6820734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69062924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACCCCCTAAATGCCTTTACTGT[C/T]CATGCTGGCTGTAAGAATGCTACTG
Phenotype
MIM: 600068
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT2B7 PubMed Links

Gene Details

Gene
UGT2B7
Gene Name
UDP glucuronosyltransferase family 2 member B7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005265702.3 Intron XP_005265759.1
XM_011532229.2 Intron XP_011530531.1
XM_011532230.1 Intron XP_011530532.1

View Full Product Details