Product Details

SNP ID
rs34324799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:167685463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGCTGAATCAATGCAAATCCAT[C/G]CCCCTACTGAAAGATGGCCTAAAGA
Phenotype
MIM: 609118
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDCD10 PubMed Links

Gene Details

Gene
PDCD10
Gene Name
programmed cell death 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007217.3 Intron NP_009148.2
NM_145859.1 Intron NP_665858.1
NM_145860.1 Intron NP_665859.1
XM_005247086.4 Intron XP_005247143.1
XM_005247087.4 Intron XP_005247144.1
XM_005247088.3 Intron XP_005247145.1
XM_006713485.3 Intron XP_006713548.1
XM_011512368.2 Intron XP_011510670.1
XM_011512369.2 Intron XP_011510671.1
XM_017005644.1 Intron XP_016861133.1
XM_017005645.1 Intron XP_016861134.1

View Full Product Details