Product Details

SNP ID
rs10513190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113231643 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTTAGGCACACCTGGTAGGAA[C/T]AGCATTGGAATGTATAATTCAAAAT
Phenotype
MIM: 603085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC31A1 PubMed Links

Gene Details

Gene
SLC31A1
Gene Name
solute carrier family 31 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001859.3 Intron NP_001850.1

View Full Product Details