Product Details

SNP ID
rs17055023
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:152427 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGAGCTGGGCCTGGGGTCAACTTCC[G/A]GCCTGGGGAAACTGGGGCAAGTATC
Phenotype
MIM: 604568
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DOC2B PubMed Links

Gene Details

Gene
DOC2B
Gene Name
double C2 domain beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003585.4 Intron NP_003576.2
XM_011535363.2 Intron XP_011533665.1
XM_017025227.1 Intron XP_016880716.1

View Full Product Details