Product Details

SNP ID
rs12542402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86214786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTGTAAAGTTGAATAAATCTG[G/T]CTTAGAAGTCAGATAAATGCCTGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC7A13 PubMed Links

Gene Details

Gene
SLC7A13
Gene Name
solute carrier family 7 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138817.2 Intron NP_620172.2
XM_011516867.1 Intron XP_011515169.1

View Full Product Details