Product Details

SNP ID
rs9656982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86214471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGTAATTGTAAATAGCAAGTC[A/G]TCTTCTCAAACCAAGCCAATCTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC7A13 PubMed Links

Gene Details

Gene
SLC7A13
Gene Name
solute carrier family 7 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138817.2 1431 Missense Mutation ACG,ATG T452M NP_620172.2
XM_011516867.1 1431 Missense Mutation ACG,ATG T443M XP_011515169.1

View Full Product Details