Product Details

SNP ID
rs33982662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136430285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGGAGAACACTGACTTGTCTCG[A/C]CCCGGCCTCACTTTCACCCGGAAGA
Phenotype
MIM: 613037 MIM: 613036 MIM: 612854
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318502.1 2236 Silent Mutation GGG,GGT G597G NP_001305431.1
NM_019892.5 2236 Silent Mutation GGG,GGT G598G NP_063945.2
XM_017014926.1 2236 Silent Mutation GGG,GGT G598G XP_016870415.1
Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
There are no transcripts associated with this gene.

Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
There are no transcripts associated with this gene.

View Full Product Details