Product Details

SNP ID
rs3737861
Assay Type
Validated
NCBI dbSNP Submissions
66
Location
Chr.1:152878462 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GCCAGACCAGGTAACACAGAGAAAC[A/C]TTGTCTTTTGGGTCCTCTCTCCATG
Phenotype
MIM: 601148
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMCP PubMed Links

Gene Details

Gene
SMCP
Gene Name
sperm mitochondria associated cysteine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030663.2 Intron NP_109588.2

View Full Product Details