Product Details

SNP ID
rs11930050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186076495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTGTATGGCATAAAACTATGA[A/G]TAATAACATCATACATGGTCATATT
Phenotype
MIM: 603029
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLR3 PubMed Links

Gene Details

Gene
TLR3
Gene Name
toll like receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003265.2 Intron NP_003256.1
XM_017008577.1 Intron XP_016864066.1

View Full Product Details