Product Details

SNP ID
rs36197001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238874016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCGGCTCTGTACCTCTCTGAGA[C/T]GGGGTTCATGCGTTAGAGGAGTTAG
Phenotype
MIM: 607556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TWIST2 PubMed Links

Gene Details

Gene
TWIST2
Gene Name
twist family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271893.3 Intron NP_001258822.1
NM_057179.2 Intron NP_476527.1

View Full Product Details