Product Details

SNP ID
rs781666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56964815 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTTCACTGACACTGTTTTTAGA[A/G]TCCTCCACAGGGCCTCACATAGATT
Phenotype
MIM: 614919
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOA1 PubMed Links

Gene Details

Gene
NOA1
Gene Name
nitric oxide associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032313.3 Intron NP_115689.1

View Full Product Details