Product Details

SNP ID
rs4659404
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:26038883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGCCCCTGCGAGTGGTAGAACA[G/T]TTGCTGAGGATTAGGCCCAAATACA
Phenotype
MIM: 601738 MIM: 609617
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXTL1 PubMed Links

Gene Details

Gene
EXTL1
Gene Name
exostosin like glycosyltransferase 1
There are no transcripts associated with this gene.

Gene
SLC30A2
Gene Name
solute carrier family 30 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004434.2 1638 UTR 3 NP_001004434.1
NM_032513.4 1638 UTR 3 NP_115902.1

View Full Product Details