Product Details
- SNP ID
-
rs4803674
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44081006 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGAGATTGAGATTGCATACGTTTG[A/T]TGTTGTAGGAGGCAGTGACCTTCAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF284
PubMed Links
Gene Details
- Gene
- ZNF284
- Gene Name
- zinc finger protein 284
View Full Product Details