Product Details

SNP ID
rs35029378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:66139817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCGTGGGTCAGGCAAATTATT[G/T]CAGGCAGTAATGCAGACTAGATGAA
Phenotype
MIM: 611037
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A26 PubMed Links

Gene Details

Gene
SLC25A26
Gene Name
solute carrier family 25 member 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164796.1 Intron NP_001158268.1
NM_173471.3 Intron NP_775742.4
XM_006712956.1 Intron XP_006713019.1
XM_011533327.2 Intron XP_011531629.1
XM_011533328.1 Intron XP_011531630.1
XM_017005671.1 Intron XP_016861160.1
XM_017005672.1 Intron XP_016861161.1
XM_017005673.1 Intron XP_016861162.1
XM_017005674.1 Intron XP_016861163.1

View Full Product Details