Product Details
- SNP ID
-
rs4327562
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:68188739 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCATCCATCCTCTTATATTTGCTTG[A/G]TGACAGTATGCATGCAGCTGTATTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FTLP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72649662] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FTLP10
- Gene Name
- ferritin light chain pseudogene 10
There are no transcripts associated with this gene.
- Gene
- TMPRSS11BNL
- Gene Name
- TMPRSS11B N-terminal like, pseudogene
There are no transcripts associated with this gene.
View Full Product Details