Product Details

SNP ID
rs12403793
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:24759461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAGCACCATCTTTTTCCTATA[C/T]AATACGTGACTTTTTTTTTTTTGTC
Phenotype
MIM: 606536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIC4 PubMed Links

Gene Details

Gene
CLIC4
Gene Name
chloride intracellular channel 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013943.2 Intron NP_039234.1

View Full Product Details