Product Details

SNP ID
rs536912
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:108971725 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTTATACAGGGATAAATTACTA[A/C]CTACCTAATTATGCTATTAGTATTT
Phenotype
MIM: 615734
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLCC1 PubMed Links

Gene Details

Gene
CLCC1
Gene Name
chloride channel CLIC like 1
There are no transcripts associated with this gene.

Gene
WDR47
Gene Name
WD repeat domain 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142550.1 Intron NP_001136022.1
NM_001142551.1 Intron NP_001136023.1
NM_014969.5 Intron NP_055784.3
XM_006710460.3 Intron XP_006710523.1
XM_011541027.2 Intron XP_011539329.1
XM_011541028.2 Intron XP_011539330.1
XM_011541029.1 Intron XP_011539331.1
XM_011541030.1 Intron XP_011539332.1
XM_011541031.1 Intron XP_011539333.1
XM_017000696.1 Intron XP_016856185.1
XM_017000697.1 Intron XP_016856186.1

View Full Product Details