Product Details
- SNP ID
-
rs367763
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:4895944 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGCGTGCCCTGCTCACTCCTTCCC[A/G]GGAATCATGACAAAGACTCCTGCCC
- Phenotype
-
MIM: 614581
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MMD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111391870] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MMD2
- Gene Name
- monocyte to macrophage differentiation associated 2
View Full Product Details