Product Details

SNP ID
rs522982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2984462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAACTTAAGAGATAGGATCACT[T/C]GCCTTGCTTTCTTTTTTTGTTGTTT
Phenotype
MIM: 612399
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TLE6 PubMed Links

Gene Details

Gene
TLE6
Gene Name
transducin like enhancer of split 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143986.1 Intron NP_001137458.1
NM_024760.2 Intron NP_079036.1
XM_005259645.2 Intron XP_005259702.1
XM_011528300.2 Intron XP_011526602.1
XM_011528301.1 Intron XP_011526603.1
XM_011528306.1 Intron XP_011526608.1

View Full Product Details