Product Details
- SNP ID
-
rs583686
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120465199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAATCTCTAAGACTCTTAAGTTA[C/T]GGGGTTTTAAAAAATATTTAATTCT
- Phenotype
-
MIM: 601562
MIM: 601943
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DYNLL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113401649] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DYNLL1
- Gene Name
- dynein light chain LC8-type 1
There are no transcripts associated with this gene.
- Gene
- GATC
- Gene Name
- glutamyl-tRNA amidotransferase subunit C
There are no transcripts associated with this gene.
- Gene
- SRSF9
- Gene Name
- serine and arginine rich splicing factor 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003769.2 |
|
Intron |
|
|
NP_003760.1 |
View Full Product Details