Product Details

SNP ID
rs751153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42621621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACAAATGTACACGAAAAATGC[C/T]AGGAGATCACCCAACTATTCCTCTC
Phenotype
MIM: 613213 MIM: 611520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5R3 PubMed Links

Gene Details

Gene
CYB5R3
Gene Name
cytochrome b5 reductase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000398.6 Intron NP_000389.1
NM_001129819.2 Intron NP_001123291.1
NM_001171660.1 Intron NP_001165131.1
NM_001171661.1 Intron NP_001165132.1
NM_007326.4 Intron NP_015565.1
Gene
POLDIP3
Gene Name
DNA polymerase delta interacting protein 3
There are no transcripts associated with this gene.

Gene
RNU12
Gene Name
RNA, U12 small nuclear
There are no transcripts associated with this gene.

View Full Product Details