Product Details
- SNP ID
-
rs635592
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138653210 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAATTTGATTCAAAATTTCCATAG[A/G]TTTCTATTTGTGGAACCCAATAAAT
- Phenotype
-
MIM: 602925
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PIK3CB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74473620] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PIK3CB
- Gene Name
- phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
View Full Product Details