Product Details
- SNP ID
-
rs583514
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:173396377 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGCTCTTCGCTGTCCCCTCCCG[C/T]TTACAGAAGCATCTGTGGTGCTTTT
- Phenotype
-
MIM: 600568
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NLGN1
PubMed Links
Gene Details
- Gene
- NLGN1
- Gene Name
- neuroligin 1
View Full Product Details