Product Details
- SNP ID
-
rs189588
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:154278066 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCACACTATACCACTGAAGAATAAA[G/T]AACAAAAAAGTACCTAAAATAATTA
- Phenotype
-
MIM: 612767
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DHX36
PubMed Links
Gene Details
- Gene
- DHX36
- Gene Name
- DEAH-box helicase 36
View Full Product Details