Product Details

SNP ID
rs515552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:100992365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACATCTCCTGGTCAGGTCACGA[G/A]AAAGAACACAGGCTTGACTATTGTC
Phenotype
MIM: 615936
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP42 PubMed Links

Gene Details

Gene
ARHGAP42
Gene Name
Rho GTPase activating protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152432.2 286 Intron NP_689645.2
XM_011542615.2 286 UTR 3 XP_011540917.1
XM_011542616.2 286 UTR 3 XP_011540918.1
XM_011542617.2 286 UTR 3 XP_011540919.1
XM_017017237.1 286 UTR 3 XP_016872726.1
XM_017017238.1 286 UTR 3 XP_016872727.1
Gene
TMEM133
Gene Name
transmembrane protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032021.2 286 Silent Mutation GAA,GAG E19E NP_114410.1

View Full Product Details