Product Details

SNP ID
rs359666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128351036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGGGAGGCTGGGGCTGCTTCC[C/T]GAGGGCCGCGGGGGACTGAGGACGC
Phenotype
MIM: 612074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRT4 PubMed Links

Gene Details

Gene
PRRT4
Gene Name
proline rich transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114726.2 1683 UTR 3 NP_001108198.2
NM_001174164.1 1683 Silent Mutation TCA,TCG S840S NP_001167635.1
XM_005250342.3 1683 Silent Mutation TCA,TCG S840S XP_005250399.1
XM_005250343.3 1683 Silent Mutation TCA,TCG S840S XP_005250400.1
XM_006715985.3 1683 Silent Mutation TCA,TCG S840S XP_006716048.1
XM_017012205.1 1683 Silent Mutation TCA,TCG S840S XP_016867694.1
XM_017012206.1 1683 UTR 3 XP_016867695.1
Gene
RBM28
Gene Name
RNA binding motif protein 28
There are no transcripts associated with this gene.

View Full Product Details