Product Details
- SNP ID
-
rs289885
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:151252645 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGACTCAGGGAGTTTGCATTTTC[C/T]TTCCTAGAAGTCATGGAGTGAGAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBM43
PubMed Links
Gene Details
- Gene
- RBM43
- Gene Name
- RNA binding motif protein 43
View Full Product Details