Product Details

SNP ID
rs612387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64944709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGCACATGTAAATGTTTACTC[G/T]ATAACTGCTCATGTTATGAGTCCTG
Phenotype
MIM: 609651
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPHA2 PubMed Links

Gene Details

Gene
GPHA2
Gene Name
glycoprotein hormone alpha 2
There are no transcripts associated with this gene.

Gene
MAJIN
Gene Name
membrane anchored junction protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037225.2 Intron NP_001032302.1
NM_001300803.1 Intron NP_001287732.1
NM_001318808.1 Intron NP_001305737.1
XM_005273918.2 Intron XP_005273975.1
XM_006718502.2 Intron XP_006718565.1
XM_006718503.3 Intron XP_006718566.1
XM_006718504.2 Intron XP_006718567.1
XM_006718505.2 Intron XP_006718568.1
XM_011544940.2 Intron XP_011543242.1
XM_011544941.2 Intron XP_011543243.1

View Full Product Details