Product Details
- SNP ID
-
rs506384
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59758143 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTCTGGGTAGATACAAAGAGAAC[A/G]TGTGGGGTTGATGGGTAGAAATGAG
- Phenotype
-
MIM: 600876
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10V2P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77442949] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10V2P
- Gene Name
- olfactory receptor family 10 subfamily V member 2 pseudogene
There are no transcripts associated with this gene.
- Gene
- STX3
- Gene Name
- syntaxin 3
View Full Product Details