Product Details
- SNP ID
-
rs523156
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:43519645 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATTCTTCTTCAGCATCTTCTGTCT[C/G]ATTTGCCACTTTCTTTCTTTGCTGA
- Phenotype
-
MIM: 600178
MIM: 605230
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MAP1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117664379] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAP1A
- Gene Name
- microtubule associated protein 1A
- Gene
- TP53BP1
- Gene Name
- tumor protein p53 binding protein 1
There are no transcripts associated with this gene.
View Full Product Details