Product Details

SNP ID
rs694985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43521189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGATCTAAGGGAAAGTCTCATA[C/T]TGCATGACCATGGGGGGCCCTGGCA
Phenotype
MIM: 600178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1A PubMed Links

Gene Details

Gene
MAP1A
Gene Name
microtubule associated protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002373.5 Intron NP_002364.5
XM_017022189.1 Intron XP_016877678.1

View Full Product Details