Product Details

SNP ID
rs426120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127517800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGAGCGCCAGGTGGCTCCGAGG[C/G]GCGTGTATGAGGGGGGTGGCATGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRRC1 PubMed Links

Gene Details

Gene
PRRC1
Gene Name
proline rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286808.1 177 Intron NP_001273737.1
NM_130809.4 177 Intron NP_570721.1
XM_006714529.3 177 UTR 5 XP_006714592.1

View Full Product Details