Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001201427.1 | 1026 | Intron | NP_001188356.1 | ||
NM_015345.3 | 1026 | Intron | NP_056160.2 | ||
XM_006715039.3 | 1026 | Intron | XP_006715102.3 | ||
XM_006715040.3 | 1026 | Intron | XP_006715103.3 | ||
XM_006715042.2 | 1026 | Intron | XP_006715105.1 | ||
XM_006715043.2 | 1026 | Intron | XP_006715106.1 | ||
XM_006715045.3 | 1026 | Intron | XP_006715108.1 | ||
XM_006715046.3 | 1026 | Intron | XP_006715109.1 | ||
XM_017010630.1 | 1026 | UTR 5 | XP_016866119.1 |