Product Details

SNP ID
rs429254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120815642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCAAAAATTTCCCTTTAAATC[A/C]TAGGCGTTGGTCATATTGAATTACT
Phenotype
MIM: 604452
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PSMD5 PubMed Links

Gene Details

Gene
PSMD5
Gene Name
proteasome 26S subunit, non-ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270427.1 1426 Intron NP_001257356.1
NM_005047.3 1426 Intron NP_005038.1
XM_011518866.2 1426 UTR 3 XP_011517168.1
XM_017014927.1 1426 Intron XP_016870416.1
XM_017014928.1 1426 Intron XP_016870417.1

View Full Product Details