Product Details

SNP ID
rs768450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:100164003 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGAAATTTATTTATATTTTTCT[A/G]GAAATGTAAGAGTGAACAGACCTAA
Phenotype
MIM: 603395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPAG1 PubMed Links

Gene Details

Gene
SPAG1
Gene Name
sperm associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003114.4 Intron NP_003105.2
NM_172218.2 Intron NP_757367.1
XM_011517240.2 Intron XP_011515542.1
XM_011517241.2 Intron XP_011515543.1
XM_011517242.2 Intron XP_011515544.1
XM_011517243.2 Intron XP_011515545.1
XM_011517245.2 Intron XP_011515547.1
XM_017013754.1 Intron XP_016869243.1
XM_017013755.1 Intron XP_016869244.1

View Full Product Details