Product Details

SNP ID
hCV721701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50003229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCACACCCAAGGCTTGCAGAA[A/G]AGCGAGCCTTCTCTTCCCTCTCCAG
Phenotype
MIM: 300008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN5 PubMed Links

Gene Details

Gene
CLCN5
Gene Name
chloride voltage-gated channel 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000084.4 Intron NP_000075.1
NM_001127898.3 Intron NP_001121370.1
NM_001127899.3 Intron NP_001121371.1
NM_001272102.1 Intron NP_001259031.1
NM_001282163.1 Intron NP_001269092.1
Gene
MIR188
Gene Name
microRNA 188
There are no transcripts associated with this gene.

Gene
MIR362
Gene Name
microRNA 362
There are no transcripts associated with this gene.

Gene
MIR500A
Gene Name
microRNA 500a
There are no transcripts associated with this gene.

Gene
MIR500B
Gene Name
microRNA 500b
There are no transcripts associated with this gene.

Gene
MIR501
Gene Name
microRNA 501
There are no transcripts associated with this gene.

Gene
MIR532
Gene Name
microRNA 532
There are no transcripts associated with this gene.

View Full Product Details