Product Details

SNP ID
rs689884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51752061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCGGGGCTGTGTTCTTCCACTC[C/T]GGGCTTTGTTCCCCCACTTTTCCCC
Phenotype
MIM: 602928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD2 PubMed Links

Gene Details

Gene
LYSMD2
Gene Name
LysM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143917.1 Intron NP_001137389.1
NM_153374.2 Intron NP_699205.1
XM_017022057.1 Intron XP_016877546.1
Gene
TMOD2
Gene Name
tropomodulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142885.1 Intron NP_001136357.1
NM_014548.3 Intron NP_055363.1
XM_017022089.1 Intron XP_016877578.1

View Full Product Details