Product Details

SNP ID
rs422767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6997424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGTGGGTGACCCGTGAGAGCA[C/G]ACAGCTAAGGGAAGAAAACATAGGA
Phenotype
MIM: 152391
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALOX12 PubMed Links

Gene Details

Gene
ALOX12
Gene Name
arachidonate 12-lipoxygenase, 12S type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000697.2 Intron NP_000688.2
XM_011523780.2 Intron XP_011522082.1
Gene
ALOX12-AS1
Gene Name
ALOX12 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details