Product Details

SNP ID
rs522837
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:9995432 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTGTCTGAGAACTTAAAAAAAACT[A/T]ATAGGACACAGGTAAAATTGTACAC
Phenotype
MIM: 612088 MIM: 606783
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLEC12A PubMed Links

Gene Details

Gene
CLEC12A
Gene Name
C-type lectin domain family 12 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207010.1 Intron NP_001193939.1
NM_001300730.1 Intron NP_001287659.1
NM_138337.5 Intron NP_612210.4
NM_201623.3 Intron NP_963917.2
XM_005253324.2 Intron XP_005253381.1
XM_006719033.3 Intron XP_006719096.1
XM_006719035.3 Intron XP_006719098.1
XM_006719036.3 Intron XP_006719099.1
XM_011520570.1 Intron XP_011518872.1
XM_011520571.2 Intron XP_011518873.2
XM_011520573.2 Intron XP_011518875.1
Gene
CLEC1B
Gene Name
C-type lectin domain family 1 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099431.1 Intron NP_001092901.1
NM_016509.3 Intron NP_057593.3
XM_005253382.4 Intron XP_005253439.1
XM_011520685.2 Intron XP_011518987.1
XM_011520686.2 Intron XP_011518988.1
XM_017019395.1 Intron XP_016874884.1
XM_017019396.1 Intron XP_016874885.1

View Full Product Details