Product Details

SNP ID
rs622289
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2665833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATAAGTTGATAACTTGTACGTT[A/G]GTATGAAATACTATGTCTTTTGTCT
Phenotype
MIM: 614982
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SMCHD1 PubMed Links

Gene Details

Gene
SMCHD1
Gene Name
structural maintenance of chromosomes flexible hinge domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015295.2 Intron NP_056110.2
XM_011525642.1 Intron XP_011523944.1
XM_011525643.2 Intron XP_011523945.1
XM_011525644.2 Intron XP_011523946.1
XM_017025684.1 Intron XP_016881173.1

View Full Product Details