Product Details

SNP ID
rs206120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32317569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCCCATGTCGCAACATCATGC[A/G]GTGATTATTTGGAAGATAGTGGTGT
Phenotype
MIM: 600185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRCA2 PubMed Links

Gene Details

Gene
BRCA2
Gene Name
BRCA2, DNA repair associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000059.3 Intron NP_000050.2
Gene
ZAR1L
Gene Name
zygote arrest 1-like
There are no transcripts associated with this gene.

View Full Product Details