Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134476.1 | 1149 | Intron | NP_001127948.1 | ||
NM_015350.2 | 1149 | Intron | NP_056165.1 | ||
XM_005270701.4 | 1149 | Intron | XP_005270758.1 | ||
XM_005270702.4 | 1149 | Intron | XP_005270759.1 | ||
XM_005270703.4 | 1149 | UTR 5 | XP_005270760.1 | ||
XM_011541143.2 | 1149 | Intron | XP_011539445.1 | ||
XM_011541144.2 | 1149 | Intron | XP_011539446.1 | ||
XM_011541145.2 | 1149 | Intron | XP_011539447.1 | ||
XM_011541146.2 | 1149 | UTR 5 | XP_011539448.1 | ||
XM_011541148.2 | 1149 | Intron | XP_011539450.1 | ||
XM_017000883.1 | 1149 | Intron | XP_016856372.1 | ||
XM_017000884.1 | 1149 | Intron | XP_016856373.1 | ||
XM_017000885.1 | 1149 | Intron | XP_016856374.1 | ||
XM_017000886.1 | 1149 | Intron | XP_016856375.1 |