Product Details
- SNP ID
-
rs542420
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:147205372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTCAGAGTTAATTAAAAGTGTGC[G/A]TATATATATATATATATATATATAT
- Phenotype
-
MIM: 604586
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
STXBP5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200859878] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STXBP5
- Gene Name
- syntaxin binding protein 5
- Gene
- STXBP5-AS1
- Gene Name
- STXBP5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details