Product Details

SNP ID
rs444923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22624054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTAATATCCAAAGGCATATATT[C/T]GTATTTACTGTGCTTTAAACATTTG
Phenotype
MIM: 613897 MIM: 602835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FANCF PubMed Links

Gene Details

Gene
FANCF
Gene Name
Fanconi anemia complementation group F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022725.3 1788 UTR 3 NP_073562.1
Gene
GAS2
Gene Name
growth arrest specific 2
There are no transcripts associated with this gene.

View Full Product Details