Product Details

SNP ID
rs745786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22553376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATTCCATCCAGTCCTGGGCAG[G/T]GGCAGAGCTTGGCTGTTTCTGGTCC
Phenotype
MIM: 610795
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SORBS3 PubMed Links

Gene Details

Gene
SORBS3
Gene Name
sorbin and SH3 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018003.2 Intron NP_001018003.1
NM_005775.4 Intron NP_005766.3

View Full Product Details