Product Details

SNP ID
rs718940
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:73063257 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCTAACAAACTGGTGGGTAGGGAT[A/G]AGAGGAGTGGCAGTGAGGGAGTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBSPON PubMed Links

Gene Details

Gene
SBSPON
Gene Name
somatomedin B and thrombospondin type 1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153225.3 5329 Intron NP_694957.3
XM_017013145.1 5329 UTR 3 XP_016868634.1

View Full Product Details