Product Details

SNP ID
rs221984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33081711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAACTCTGAAGTACCCAGGAAC[C/T]TCCTCCTGCTAGCAGAGTTCACATC
Phenotype
MIM: 615717 MIM: 615718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPIFB3 PubMed Links

Gene Details

Gene
BPIFB3
Gene Name
BPI fold containing family B member 3
There are no transcripts associated with this gene.

Gene
BPIFB4
Gene Name
BPI fold containing family B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182519.2 Intron NP_872325.2

View Full Product Details