Product Details

SNP ID
rs762105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102363838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGGGTCAGTGCATGTAAAGTTA[A/G]GCACGGTGCCTGCCGCTGTACATTT
Phenotype
MIM: 613362 MIM: 615000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CINP PubMed Links

Gene Details

Gene
CINP
Gene Name
cyclin dependent kinase 2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320046.1 Intron NP_001306975.1
NM_032630.2 Intron NP_116019.1
Gene
TECPR2
Gene Name
tectonin beta-propeller repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172631.2 Intron NP_001166102.1
NM_014844.4 Intron NP_055659.2

View Full Product Details