Product Details

SNP ID
rs640907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74332205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGCTTTGGCTCCCCAAGTACA[A/G]GGAAACCTGAAATAGAATGTTATGT
Phenotype
MIM: 611610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928580 PubMed Links

Gene Details

Gene
LOC101928580
Gene Name
uncharacterized LOC101928580
There are no transcripts associated with this gene.

Gene
PGM2L1
Gene Name
phosphoglucomutase 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173582.4 6675 UTR 3 NP_775853.2
XM_011544953.2 6675 UTR 3 XP_011543255.1

View Full Product Details