Product Details

SNP ID
rs13306575
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:183563302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGCTCATTGCTGTCCCGAGGCC[A/G]ATAGCTGGGTGGGGATAATGAGTAA
Phenotype
MIM: 608515 MIM: 610964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCF2 PubMed Links

Gene Details

Gene
NCF2
Gene Name
neutrophil cytosolic factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000433.3 1458 Silent Mutation AGG,GGG R395G NP_000424.2
NM_001127651.2 1458 Silent Mutation AGG,GGG R395G NP_001121123.1
NM_001190789.1 1458 Silent Mutation AGG,GGG R314G NP_001177718.1
NM_001190794.1 1458 Silent Mutation AGG,GGG R350G NP_001177723.1
XM_005245207.1 1458 Silent Mutation AGG,GGG R359G XP_005245264.1
XM_011509580.1 1458 Missense Mutation AGG,GGG R395G XP_011507882.1
XM_011509581.1 1458 Silent Mutation AGG,GGG R395G XP_011507883.1
Gene
SMG7
Gene Name
SMG7, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details